2-Iodotoluene ≥98.0% stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with Stabilised

I0217-25G I0217-250G
TCI0217-025GEA 64.94 CAD
TCI0217-025G TCI0217-250G
2-Iodotoluene ≥98.0% stabilized
2-Iodotoluene

(stabilized with Copper chip)
CAS Number: 615-37-2
MDL Number: MFCD00001042
Molecular Formula: C7H7I
Molecular Weight: 218.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Flash Point (°C): 90
Specific Gravity (20/20): 1.72

Formula: C₇H₇I
Boiling Pt: 211 °C
Density: 1.72 (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001042
CAS Number: 615-37-2
EINECS: 210-422-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc5ae225e155a074698efd1e560b1a1664063433&vid=cfaf3693b5a746301ce6f33c51dab52b1a421433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR