3-Iodotoluene ≥98.0%

Supplier: TCI America
Warning

I0216-500G I0216-25G
TCI0216-500GEA 559.32 CAD
TCI0216-500G TCI0216-025G
3-Iodotoluene ≥98.0%
3-Iodotoluene

CAS Number: 625-95-6
MDL Number: MFCD00001050
Molecular Formula: C7H7I
Molecular Weight: 218.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 213
Melting point (°C): -27
Flash Point (°C): 82
Specific Gravity (20/20): 1.70

Formula: C₇H₇I
Boiling Pt: 213 °C
Melting Pt: –27 °C
Density: 1.70 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00001050
CAS Number: 625-95-6
EINECS: 210-918-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=827bfda778bb95e4620ba4710f402c69a674ac0c&vid=8fc6cfcae7e24f898813daa857800e56497ccfef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR