L(+)-Isoleucine ≥98.0%, crystals

Supplier: TCI America

Synonyms: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH

I0181-25G I0181-500G I0181-100G
TCI0181-025GEA 82.37 CAD
TCI0181-025G TCI0181-500G TCI0181-100G
L(+)-Isoleucine ≥98.0%, crystals
L(+)-Isoleucine

CAS Number: 73-32-5 MDL Number: MFCD00064222 Molecular Formula: C6H13NO2 Molecular Weight: 131.18 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 288 Specific rotation [a]20/D: 40.5 deg (C=4, 6mol/L HCl)


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Ile-OH

Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Melting Pt: 288 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bceb3b99593bc0ea02f9ee279d1600fc4ad4e9b2&vid=948f14df5f68c9caf589185b29e22fc56e8d7842&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR