Dimethyl isophthalate ≥99.0%

Supplier: TCI America

I0157-25G I0157-500G
TCI0157-025GEA 30.56 CAD
TCI0157-025G TCI0157-500G
Dimethyl isophthalate ≥99.0%
Dimethyl isophthalate

CAS Number: 1459-93-4
MDL Number: MFCD00008433
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 124
Melting point (°C): 69
Flash Point (°C): 173


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Isophthalic Acid Dimethyl Ester

Formula: C₁₀H₁₀O₄
Boiling Pt: 124 °C
Melting Pt: 69 °C
Density: 1.186 g/cm³ (25 °C)
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00008433
CAS Number: 1459-93-4
EINECS: 215-951-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cd642a8241d6263620a88d10ccb3283b893dffb&vid=5cd642a8241d6263620a88d10ccb3283b893dffb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR