Isophthalic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

I0155-25G I0155-500G
TCI0155-025GEA 30.02 CAD
TCI0155-025G TCI0155-500G
Isophthalic acid ≥99.0% (by GC, titration analysis)
Isophthalic acid

CAS Number: 121-91-5
MDL Number: MFCD00002516
Molecular Formula: C8H6O4
Molecular Weight: 166.13
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 340


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers
Other Product Names / Synonyms: m-Phthalic Acid, Benzene-1,3-dicarboxylic Acid

Formula: C₈H₆O₄
Boiling Pt: 119…121 °C (6 mmHg)
Melting Pt: 340 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002516
CAS Number: 121-91-5
EINECS: 204-506-4
Merck Index: 13,05216
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64c16d776c99f84bcae02c2b516c64534b3934f7&vid=64c16d776c99f84bcae02c2b516c64534b3934f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR