Isophorone ≥97.0%
Supplier: TCI America
|
Warning
|
Synonyms:
3,5,5-Trimethyl-2-cyclohexen-1-one, 3,5,5-Trimethylcyclohex-2-enone
I0151-500ML
I0151-25ML
TCI0151-500MLEA
34.76
CAD
TCI0151-500ML
TCI0151-25ML
Isophorone ≥97.0%
Isophorone
CAS Number: 78-59-1
MDL Number: MFCD00001584
Molecular Formula: C9H14O
Molecular Weight: 138.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Melting point (°C): -8
Flash Point (°C): 84
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: 3,5,5-Trimethyl-2-cyclohexen-1-one
Formula:
C₉H₁₄O Boiling Pt: 215 °C Melting Pt: –8 °C Density: 0.92 (20 °C) Flash Pt: 84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001584 CAS Number: 78-59-1 EINECS: 201-126-0 Merck Index: 13,05215 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...