Ethyl isonicotinate ≥98.0%
Supplier: TCI America
Synonyms:
4-Picolinic acid ethyl ester, 4-Pyridinecarboxylic acid ethyl ester, Pyridine-4-carboxylic acid ethyl ester, Ethyl 4-pyridinecarboxylate, Isonicotinic acid ethyl ester
CAS Number: 1570-45-2
MDL Number: MFCD00006428
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 87
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: Ethyl 4-Pyridinecarboxylate, Isonicotinic Acid Ethyl Ester
Formula:
C₈H₉NO₂ Boiling Pt: 92 °C Density: 1.11 (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00006428 CAS Number: 1570-45-2 EINECS: 216-379-2 Merck Index: 13,05206 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...