Isonicotinamide ≥99.0%

Supplier: TCI America

Synonyms: 4-Pyridinecarboxylic acid amide

I0135-25G I0135-500G
TCI0135-025GEA 47.76 CAD
TCI0135-025G TCI0135-500G
Isonicotinamide ≥99.0%
Isonicotinamide

CAS Number: 1453-82-3
MDL Number: MFCD00006432
Molecular Formula: C6H6N2O
Molecular Weight: 122.13
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 158

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 334
Melting Pt: 158 °C
MDL Number: MFCD00006432
CAS Number: 1453-82-3
EINECS: 215-926-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b03b9ab21c43a22a372f95b74275cafc59ff527&vid=3b03b9ab21c43a22a372f95b74275cafc59ff527&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR