Isoeugenol ≥97.0%

Supplier: TCI America
Warning

Synonyms: Propenylguaiacol, 4-Hydroxy-3-methoxy-1-propenylbenzene, 2-Methoxy-4-(prop-1-en-1-yl)phenol

I0132-500G I0132-25G
TCI0132-500GEA 303.36 CAD
TCI0132-500G TCI0132-025G
Isoeugenol ≥97.0%
Isoeugenol

CAS Number: 97-54-1
MDL Number: MFCD00009285
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 134
Specific Gravity (20/20): 1.08


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: 4-Hydroxy-3-methoxy-1-propenylbenzene, Propenylguaiacol

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.08 (20 °C)
Flash Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00009285
CAS Number: 97-54-1
EINECS: 202-590-7
Merck Index: 13,05189
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f50afb99fb51c121de84ef3b598d32ee1a3fe71&vid=e63646e8e7bb6520054730f1f1b6e2412fddf927&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR