Isobutyrophenone ≥98.0%

Supplier: TCI America

I0113-25ML I0113-500ML
TCI0113-025MLEA 69.36 CAD
TCI0113-025ML TCI0113-500ML
Isobutyrophenone ≥98.0%
Isobutyrophenone

CAS Number: 611-70-1
MDL Number: MFCD00008917
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 84
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Isopropyl Phenyl Ketone

Formula: (C₆H₅)C(O)CH(CH₃)₂
Boiling Pt: 215 °C
Melting Pt: 1 °C
Density: 0.98 (20 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008917
CAS Number: 611-70-1
EINECS: 210-275-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da46257bdd61190d1f1eaebd21b0ac320c0689c6&vid=dba56279428dcc6950f50010ead0b0d16f15258c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR