Isatin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0080-25G I0080-500G I0080-100G
TCI0080-025GEA 63.66 CAD
TCI0080-025G TCI0080-500G TCI0080-100G
Isatin ≥98.0% (by GC, titration analysis)
Isatin

CAS Number: 91-56-5
MDL Number: MFCD00005718
Molecular Formula: C8H5NO2
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 203


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: Indole-2,3-dione, 2,3-Indolinedione

Formula: C₈H₅NO₂
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 203 °C
Density: 1.472 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00005718
CAS Number: 91-56-5
EINECS: 202-077-8
Merck Index: 13,05119
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5f20ce6e460f08a90897432ba5c63e36e31461&vid=4d5f20ce6e460f08a90897432ba5c63e36e31461&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR