Iron(III) acetylacetonate ≥98.0%

Supplier: TCI America
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

I0079-25G I0079-500G I0079-100G
TCI0079-025GEA 41.43 CAD
TCI0079-025G TCI0079-500G TCI0079-100G
Iron(III) acetylacetonate ≥98.0%
Iron (III) acetylacetonate

CAS Number: 14024-18-1
MDL Number: MFCD00000020
Molecular Formula: C15H21FeO6
Molecular Weight: 353.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 185


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Oxidation
Other Product Names / Synonyms: Acetylacetone Iron(III) Salt, Ferric(III) Acetylacetonate, Iron(III) Acetylacetonate

Formula: C₁₅H₂₁FeO₆
MW: 353.18 g/mol
Boiling Pt: 100
Melting Pt: 185 °C
Density: 5.24 g/cm³ (20 °C)
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d417943775d10556d56627005a2062df8d24c5d&vid=4d417943775d10556d56627005a2062df8d24c5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR