α-Ionone ≥90.0%

Supplier: TCI America

Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

I0076-25G I0076-250G
TCI0076-025GEA 140.62 CAD
TCI0076-025G TCI0076-250G
α-Ionone ≥90.0%
α-Ionone

CAS Number: 127-41-3
MDL Number: MFCD00001565
Molecular Formula: C13H20O
Molecular Weight: 192.30
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 104
Specific Gravity (20/20): 0.93


Product Categories: Bioscience, Terpenes (Others)
Other Product Names / Synonyms: alpha-Jonone

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Boiling Pt: 148 °C
Density: 0.93 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00001565
CAS Number: 127-41-3
EINECS: 204-841-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4d4ecf1ae7fb50e5bca83cfb1eb1a7921475e67&vid=f4d4ecf1ae7fb50e5bca83cfb1eb1a7921475e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR