N-Iodosuccinimide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
I0074-25G
I0074-100G
I0074-5G
TCI0074-025GEA
221.52
CAD
TCI0074-025G
TCI0074-100G
TCI0074-005G
N-Iodosuccinimide ≥98.0% (by titrimetric analysis)
N-Iodosuccinimide
CAS Number: 516-12-1
MDL Number: MFCD00005512
Molecular Formula: C4H4INO2
Molecular Weight: 224.99
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 201
Specific rotation [a]20/D: -2 deg (C=1, CHCl3)
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Oxidation, Halogenation, Glycosylation, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: NIS
Formula:
C₄H₄INO₂ Boiling Pt: 356 °C (1013 hPa) Melting Pt: 201 °C Density: 2.245 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005512 CAS Number: 516-12-1 EINECS: 208-221-6 Merck Index: 13,05065 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...