1-Iodo-4-nitrobenzene ≥99.0%

Supplier: TCI America

I0064-25G I0064-100G
TCI0064-025GEA 90.06 CAD
TCI0064-025G 76219-982
1-Iodo-4-nitrobenzene ≥99.0%
1-Iodo-4-nitrobenzene

CAS Number: 636-98-6
MDL Number: MFCD00007299
Molecular Formula: C6H4INO2
Molecular Weight: 249.01
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: 4-Iodonitrobenzene

Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 173 °C
Density: 2.273 g/cm³ (20 °C)
MDL Number: MFCD00007299
CAS Number: 636-98-6
EINECS: 211-272-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e7a2d2592f6100c4ddecea46b1aca194639049d&vid=5e7a2d2592f6100c4ddecea46b1aca194639049d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR