Iodocyclohexane ≥98.0% stabilised

Supplier: TCI America

Synonyms: Cyclohexyl iodide

I0056-100G I0056-10G
TCI0056-100GEA 477.63 CAD
TCI0056-100G TCI0056-010G
Iodocyclohexane ≥98.0% stabilised
Iodocyclohexane

(stabilized with Copper chip)
CAS Number: 626-62-0
MDL Number: MFCD00003826
Molecular Formula: C6H11I
Molecular Weight: 210.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 72
Specific Gravity (20/20): 1.63


Other Product Names / Synonyms: Cyclohexyl Iodide

Formula: C₆H₁₁I
MW: 210.06 g/mol
Boiling Pt: 80
Density: 1.63 (20 °C)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003826
CAS Number: 626-62-0
EINECS: 210-957-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47219e3f7b60584b110b88338065a0f641539e52&vid=47219e3f7b60584b110b88338065a0f641539e52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR