4-Iodobenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: p-Iodobenzoic acid

I0054-25G I0054-5G
TCI0054-025GEA 76 CAD
TCI0054-025G TCI0054-5G
4-Iodobenzoic acid ≥98.0%
4-Iodobenzoic acid

CAS Number: 619-58-9
MDL Number: MFCD00002533
Molecular Formula: C7H5IO2
Molecular Weight: 248.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 319
Density: 2.184 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002533
CAS Number: 619-58-9
EINECS: 210-603-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8d4df5d48e8676d65c091cd27205b7792b13671&vid=c8d4df5d48e8676d65c091cd27205b7792b13671&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR