2-Iodobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: o-Iodobenzoic acid

I0053-500G I0053-25G I0053-100G
TCI0053-500GEA 398.16 CAD
TCI0053-500G TCI0053-025G TCI0053-100G
2-Iodobenzoic acid ≥98.0% (by GC, titration analysis)
2-Iodobenzoic acid

CAS Number: 88-67-5
MDL Number: MFCD00002419
Molecular Formula: C7H5IO2
Molecular Weight: 248.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 162
Flash Point (°C): 110

Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 162 °C
Density: 2.25 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002419
CAS Number: 88-67-5
EINECS: 201-850-7
Merck Index: 13,05052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4d1615214cf4420f881d571deffaca7a94b180f&vid=5d42793d9eabba328b75b5e270444fcc42c04a33&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR