3-Iodobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0052-5G I0052-25G
TCI0052-5GEA 44.24 CAD
TCI0052-5G TCI0052-025G
3-Iodobenzoic acid ≥98.0% (by GC, titration analysis)

CAS Number: 618-51-9
MDL Number: MFCD00002496
Molecular Formula: C7H5IO2
Molecular Weight: 248.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 187

Formula: C₇H₅IO₂
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 187 °C
Density: 2.171 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002496
CAS Number: 618-51-9
EINECS: 210-555-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd22f00c02f374cde694dff67ca49a01d7f3f466&vid=b03fe9225d4e27b17e6fef0273d498c9b56a995a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR