Iodobenzene ≥99.0%

Supplier: TCI America
Warning

Synonyms: Phenyliodide

I0050-25G I0050-500G I0050-100G
TCI0050-025GEA 57.97 CAD
TCI0050-025G TCI0050-500G TCI0050-100G
Iodobenzene ≥99.0%
Iodobenzene

CAS Number: 591-50-4
MDL Number: MFCD00001029
Molecular Formula: C6H5I
Molecular Weight: 204.01
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Melting point (°C): -29
Flash Point (°C): 74
Specific Gravity (20/20): 1.83

Formula: C₆H₅I
MW: 204.01 g/mol
Boiling Pt: 188 °C
Melting Pt: –29 °C
Density: 1.83 (20 °C)
Flash Pt: 74
Storage Temperature: Ambient
MDL Number: MFCD00001029
CAS Number: 591-50-4
EINECS: 209-719-6
Merck Index: 13,05051
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97c144a3cd20c73544b4ee4c31536141d065680a&vid=cb8a7cd5aca6d2e369927b47f5b618087aab2483&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR