3-Iodoaniline ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I0047-5G I0047-25G
TCI0047-5GEA 41.08 CAD
TCI0047-5G TCI0047-025G
3-Iodoaniline ≥99.0% (by GC, titration analysis)
3-Iodoaniline

CAS Number: 626-01-7
MDL Number: MFCD00007781
Molecular Formula: C6H6IN
Molecular Weight: 219.03
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 116
Flash Point (°C): 113
Freezing point (°C): 25

Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 116 °C
Melting Pt: 24…26 °C
Density: 1.821 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007781
CAS Number: 626-01-7
EINECS: 210-922-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c1dd413512c4d08189d9af66f9d61a4a0d5e8ed&vid=36bd64ea2633e9060f885099ca28d57990d2b29e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR