2-Iodoaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I0046-5G I0046-25G
TCI0046-005GEA 58.46 CAD
TCI0046-005G TCI0046-025G
2-Iodoaniline ≥98.0% (by GC, titration analysis)
2-Iodoaniline

CAS Number: 615-43-0
MDL Number: MFCD00007680
Molecular Formula: C6H6IN
Molecular Weight: 219.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 57
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₆H₆IN
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 57 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007680
CAS Number: 615-43-0
EINECS: 210-426-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff530ada4776ad57a4996edb867b2d649644bac5&vid=2b0f660c3448471ee9529af77dd6de9aab78ccfc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR