Inosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0037-500G I0037-25G
TCI0037-500GEA 1000.57 CAD
TCI0037-500G TCI0037-025G
Inosine ≥98.0% (by HPLC, titration analysis)
Inosine

CAS Number: 58-63-9
MDL Number: MFCD00066770
Molecular Formula: C10H12N4O5
Molecular Weight: 268.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 226
Specific rotation [a]20/D: -52 deg (C=1, H2O)

Formula: C₁₀H₁₂N₄O₅
Melting Pt: 226 °C
Storage Temperature: Ambient
MDL Number: MFCD00066770
CAS Number: 58-63-9
EINECS: 200-390-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5365f011d06506e0b6f6e5ca6b77394d7ac3d9b8&vid=5365f011d06506e0b6f6e5ca6b77394d7ac3d9b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR