3-(2-Hydroxyethyl)indole ≥98.0%
Supplier: TCI America
I0030-5G
I0030-25G
I0030-1G
TCI0030-5GEA
188.02
CAD
TCI0030-5G
TCI0030-25G
TCI0030-001G
3-(2-Hydroxyethyl)indole ≥98.0%
3-(2-Hydroxyethyl)indole
CAS Number: 526-55-6
MDL Number: MFCD00005659
Molecular Formula: C10H11NO
Molecular Weight: 161.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 179
Melting point (°C): 59
Other Product Names / Synonyms: 3-(2-Hydroxyethyl)indole, Tryptophol
Formula:
C₁₀H₁₁NO Boiling Pt: 179 °C Melting Pt: 59 °C |
MDL Number:
MFCD00005659 CAS Number: 526-55-6 EINECS: 208-393-2 Merck Index: 12,09930 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...