Indole-5-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0029-1G I0029-5G
TCI0029-001GEA 67.94 CAD
TCI0029-001G TCI0029-5G
Indole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
Indole-5-carboxylic acid

CAS Number: 1670-81-1
MDL Number: MFCD00005678
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 212

Formula: C₉H₇NO₂
Melting Pt: 212 °C
Storage Temperature: Freezer
MDL Number: MFCD00005678
CAS Number: 1670-81-1
EINECS: 216-799-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ecd479462ebead7502e0a9b4731b51403c7d552&vid=2ecd479462ebead7502e0a9b4731b51403c7d552&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR