Indole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
I0028-5G
I0028-25G
TCI0028-005GEA
86.9
CAD
TCI0028-005G
TCI0028-25G
Indole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Indole-3-carboxylic acid
CAS Number: 771-50-6
MDL Number: MFCD00005624
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 234
Formula:
C₉H₇NO₂ Melting Pt: 234 °C |
MDL Number:
MFCD00005624 CAS Number: 771-50-6 EINECS: 212-231-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...