Indole-3-carbaldehyd ≥98.0%
Supplier: TCI America
I0027-5G
I0027-25G
TCI0027-005GEA
57.88
CAD
TCI0027-005G
TCI0027-025G
Indole-3-carbaldehyd ≥98.0%
Formula:
C₉H₇NO Melting Pt: 195 °C Density: 1.34 g/cm³ (20 °C) Flash Pt: 240 °C |
MDL Number:
MFCD00005622 CAS Number: 487-89-8 EINECS: 207-665-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...