Indol-3-acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

I0022-25G I0022-5G
TCI0022-025GEA 128.17 CAD
TCI0022-025G TCI0022-005G
Indol-3-acetic acid ≥98.0% (by titrimetric analysis)
Indol-3-acetic acid

CAS Number: 87-51-4
MDL Number: MFCD00005636
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 168


Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: IAA

Formula: C₁₀H₉NO₂
Boiling Pt: 120…125 °C (0.05 mmHg)
Melting Pt: 168 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005636
CAS Number: 87-51-4
EINECS: 201-748-2
Merck Index: 13,04986
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=890c3756018884834d6ef23cd68d83fd25db4919&vid=ea92b80bc98698961996053ae1ddd829b7119fd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR