Ninhydrin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate2,2-Dihydroxyindane-1,3-dione1,2,3-Indantrione monohydrate

I0015-500G I0015-25G I0015-100G
TCI0015-100GEA 350.87 CAD
TCI0015-100G TCI0015-025G TCI0015-500G
Ninhydrin ≥98.0% (by titrimetric analysis)
Ninhydrin

CAS Number: 485-47-2
MDL Number: MFCD00003791
Molecular Formula: C9H6O4
Molecular Weight: 178.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 250


Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 2,2-Dihydroxy-1,3-indandione, 1,2,3-Indantrione Monohydrate, 1,2,3-Triketohydrindene Monohydrate

Formula: C₉H₆O₄
MW: 178.14 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250 °C
Density: 0.862 g/cm³ (20 °C)
MDL Number: MFCD00003791
CAS Number: 485-47-2
Merck Index: 13,06582
https://ca.vwr.com/stibo/web/std.lang.all/18/05/28971805.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR