1-Indanone ≥98.0%

Supplier: TCI America

Synonyms: 2,3-Dihydro-1H-inden-1-one

I0013-25G I0013-5G
TCI0013-025GEA 108.61 CAD
TCI0013-025G TCI0013-5G
1-Indanone ≥98.0%
1-Indanone

CAS Number: 83-33-0
MDL Number: MFCD00003785
Molecular Formula: C9H8O
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 245
Melting point (°C): 41
Flash Point (°C): 110


Other Product Names / Synonyms: alpha-Hydrindone, 1-Hydroindone, 1-Ketohydrindene

Formula: C₉H₈O
Boiling Pt: 245 °C
Melting Pt: 41 °C
Density: 1.1028 g/cm³ (40 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003785
CAS Number: 83-33-0
EINECS: 201-470-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9be235144e8d23dc201ea5c6dab45493310b1e6a&vid=9be235144e8d23dc201ea5c6dab45493310b1e6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR