Diethanolamine ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol

I0008-25G I0008-500G
TCI0008-25GEA 36.88 CAD
TCI0008-25G TCI0008-500G
Diethanolamine ≥99.0% (by GC, titration analysis)
Diethanolamine

CAS Number: 111-42-2
MDL Number: MFCD00002843
Molecular Formula: C4H11NO2
Molecular Weight: 105.14
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 269
Flash Point (°C): 160
Freezing point (°C): 27


Other Product Names / Synonyms: 2,2'-Dihydroxydiethylamine, 2,2'-Iminodiethanol

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 269 °C
Melting Pt: 28 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a164c3a3ae3e0614f0e2d84ef586a4742ca29bca&vid=a164c3a3ae3e0614f0e2d84ef586a4742ca29bca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR