Iminodiacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

I0006-500G I0006-25G
TCI0006-500GEA 88.31 CAD
TCI0006-500G TCI0006-025G
Iminodiacetic acid ≥98.0% (by titrimetric analysis)
Iminodiacetic acid

CAS Number: 142-73-4
MDL Number: MFCD00004280
Molecular Formula: C4H7NO4
Molecular Weight: 133.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 250

Formula: C₄H₇NO₄
Melting Pt: 250 °C
MDL Number: MFCD00004280
CAS Number: 142-73-4
EINECS: 205-555-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=231836272d31b287c91de56bfd89d6e7ea324d69&vid=231836272d31b287c91de56bfd89d6e7ea324d69&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR