Imidazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Glyoxaline
, 1H-Imidazole
, Glyoxalamine
CAS Number: 288-32-4
MDL Number: MFCD00005183
Molecular Formula: C3H4N2
Molecular Weight: 68.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 90
Flash Point (°C): 145
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Glyoxaline
Formula:
C₃H₄N₂ MW: 68.08 g/mol Boiling Pt: 256 °C Melting Pt: 90 °C Density: 1.18 g/cm³ (20 °C) Flash Pt: 145 °C |
MDL Number:
MFCD00005183 CAS Number: 288-32-4 UN: 2923 ADR: 8,III Merck Index: 13,04935 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...