cis-4-Hydroxy-L-(-)-proline ≥97.0%

Supplier: TCI America

Synonyms: L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline

H1169-1G H1169-100MG
TCH1169-1GEA 486.01 CAD
TCH1169-1G TCH1169-100MG
cis-4-Hydroxy-L-(-)-proline ≥97.0%
cis-4-Hydroxy-L(-)-proline

CAS Number: 618-27-9
Molecular Formula: C5H9NO3
Molecular Weight: 131.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 241
Specific rotation [a]20/D: -59 deg (C=2, H2O)


Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic Acid, H-cis-Hyp-OH

Formula: C₅H₉NO₃
MW: 131.13 g/mol
Melting Pt: 257
MDL Number: MFCD00064319
CAS Number: 618-27-9
EINECS: 210-542-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a434aee2f6adc2bec9e82e809c81e830b88bd643&vid=a434aee2f6adc2bec9e82e809c81e830b88bd643&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR