(2-Hydroxylpropyl)-β-cyclodextrine

Supplier: TCI America

Synonyms: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

H0979-25G H0979-100G
TCH0979-025GEA 300.64 CAD
TCH0979-025G TCH0979-100G
(2-Hydroxylpropyl)-β-cyclodextrine
(2-Hydroxylpropyl)-β-cyclodextrine

CAS Number: 128446-35-5
MDL Number: MFCD00074978
Form: Crystal
Color: White
Specific rotation [a]20/D: 125 deg (C=1, H2O)


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Oligosaccharides, Cyclodextrins

Formula: C₆₃H₁₁₂O₄₂
Storage Temperature: Ambient
MDL Number: MFCD00069372
CAS Number: 128446-35-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42bcb7949075f595c5946d961028d276b97d5d4d&vid=42bcb7949075f595c5946d961028d276b97d5d4d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR