(S,S)-(+)-2,5-Hexanediol ≥98.0%
Supplier: TCI America
Synonyms:
(2S,5S)-2,5-Hexanediol, (2S,5S)-Hexane-2,5-diol, (2S,5S)-(+)-2,5-Hexanediol, (S,S)-(+)-2,5-Dihydroxy hexane
CAS Number: 34338-96-0
MDL Number: MFCD00082583
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 71
Melting point (°C): 54
Flash Point (°C): 102
Specific rotation [a]20/D: 25 deg (C=1, EtOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2S,5S)-2,5-Dihydroxyhexane
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol Boiling Pt: 71 °C Melting Pt: 54 °C Flash Pt: 102 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00082583 CAS Number: 34338-96-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...