6-Methyl-2-pyridone ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Hydroxy-6-methylpyridine, 6-Methylpyridin-2-ol, 6-Methylpyridin-2(1H)-one

H0942-5G H0942-25G
TCH0942-005GEA 46.29 CAD
TCH0942-005G TCH0942-025G
6-Methyl-2-pyridone ≥98.0%
6-Methyl-2-pyridone

CAS Number: 3279-76-3
MDL Number: MFCD00006279
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: 6-Hydroxy-2-picoline, 6-Methyl-2-pyridinol, 6-Methyl-2(1H)-pyridinone

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 283…284 °C (1013 hPa)
Melting Pt: 159 °C
Density: 1.13 g/cm³ (20 °C)
MDL Number: MFCD00006279
CAS Number: 3279-76-3
EINECS: 221-919-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c37288b09391a2862bbbc0f2bab6b15144f71260&vid=c37288b09391a2862bbbc0f2bab6b15144f71260&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR