2-Acetyl-1-naphthol ≥98.0%

Supplier: TCI America

H0935-100G H0935-25G
TCH0935-100GEA 982.76 CAD
TCH0935-100G TCH0935-025G
2-Acetyl-1-naphthol ≥98.0%
2-Acetyl-1-naphthol

CAS Number: 711-79-5
MDL Number: MFCD00003963
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 99


Other Product Names / Synonyms: 2-Acetyl-1-hydroxynaphthalene, 2-Acetyl-1-naphthol

Formula: C₁₂H₁₀O₂
Boiling Pt: 110…115 °C (0.5 mmHg)
Melting Pt: 99 °C
MDL Number: MFCD00003963
CAS Number: 711-79-5
EINECS: 211-918-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c9fca17b27f8d334c7238d6976a63df5c237590&vid=5c9fca17b27f8d334c7238d6976a63df5c237590&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR