3-Hydroxy-p-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H0929-5G H0929-25G
TCH0929-005GEA 58.46 CAD
TCH0929-005G TCH0929-025G
3-Hydroxy-p-toluic acid ≥98.0% (by GC, titration analysis)
3-Hydroxy-p-toluic acid

CAS Number: 586-30-1
MDL Number: MFCD00002511
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 211


Other Product Names / Synonyms: 3-Hydroxy-p-toluic Acid

Formula: C₈H₈O₃
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 211 °C
Storage Temperature: Ambient
MDL Number: MFCD00002511
CAS Number: 586-30-1
EINECS: 209-571-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=984d440b23900484cff0ff66e65901c8489943cf&vid=4abd8a2c26e5d6796ae2c1ab58b42d235beedda3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR