2,4-Dihydroxy-3-nitro-6-picoline ≥98.0%
Supplier: TCI America
Synonyms:
2,4-Dihydroxy-6-methyl-3-nitropyridine, 4-Hydroxy-6-methyl-3-nitropyridin-2(1H)-one, 4-Hydroxy-6-methyl-3-nitro-2-pyridone
H0921-5G
H0921-25G
TCH0921-005GEA
64.78
CAD
TCH0921-005G
TCH0921-025G
2,4-Dihydroxy-3-nitro-6-picoline ≥98.0%
2,4-Dihydroxy-3-nitro-6-picoline
CAS Number: 4966-90-9
MDL Number: MFCD00167612
Molecular Formula: C6H6N2O4
Molecular Weight: 170.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2,4-Dihydroxy-6-methyl-3-nitropyridine
Formula:
C₆H₆N₂O₄ MW: 170.12 g/mol |
MDL Number:
MFCD00167612 CAS Number: 4966-90-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...