3-Hydroxybenzyl alcohol ≥99.0%
Supplier: TCI America
H0918-25G
H0918-5G
TCH0918-25GEA
262.28
CAD
TCH0918-25G
TCH0918-005G
3-Hydroxybenzyl alcohol ≥99.0%
3-Hydroxybenzyl alcohol
CAS Number: 620-24-6
MDL Number: MFCD00004643
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 123
Melting point (°C): 72
Flash Point (°C): 73
Other Product Names / Synonyms: 3-Methylolphenol
Formula:
C₇H₈O₂ Boiling Pt: 123 °C Melting Pt: 72 °C Density: 1.161 g/cm³ (25 °C) Flash Pt: 73 °C |
MDL Number:
MFCD00004643 CAS Number: 620-24-6 EINECS: 210-633-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...