1-Hexylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Hexylboronic acid

H0913-1G H0913-5G
TCH0913-001GEA 34.76 CAD
TCH0913-001G TCH0913-005G
1-Hexylboronic acid (contains varying amounts of Anhydride)
1-Hexylboronic acid

CAS Number: 16343-08-1
MDL Number: MFCD01074641
Molecular Formula: C6H15BO2
Molecular Weight: 129.99
Form: Crystal
Color: White


Other Product Names / Synonyms: 1-Hexaneboronic Acid

Formula: C₆H₁₅BO₂
MDL Number: MFCD01074641
CAS Number: 16343-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9365490736db6f59fe79c655d618c01d2ebda4fd&vid=9365490736db6f59fe79c655d618c01d2ebda4fd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR