4-Hydroxy-3-nitrobenzoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
H0910-25G
H0910-100G
TCH0910-025GEA
58.46
CAD
TCH0910-025G
TCH0910-100G
4-Hydroxy-3-nitrobenzoic acid ≥98.0%
4-Hydroxy-3-nitrobenzoic acid
CAS Number: 616-82-0
MDL Number: MFCD00007119
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 184
Formula:
C₇H₅NO₅ MW: 183.12 g/mol Melting Pt: 184 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007119 CAS Number: 616-82-0 EINECS: 210-494-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...