1-(4-Hydroxyphenyl)piperazine ≥98.0%

Supplier: TCI America

H0891-250G H0891-25G
TCH0891-250GEA 919.09 CAD
TCH0891-250G TCH0891-025G
1-(4-Hydroxyphenyl)piperazine ≥98.0%
1-(4-Hydroxyphenyl)piperazine

CAS Number: 56621-48-8
MDL Number: MFCD00066156
Molecular Formula: C10H14N2O
Molecular Weight: 178.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 223


Other Product Names / Synonyms: 4-(1-Piperazinyl)phenol

Formula: C₁₀H₁₄N₂O
Melting Pt: 223 °C
MDL Number: MFCD00066156
CAS Number: 56621-48-8
EINECS: 260-289-6
UN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17e0b58af541c85e89fb66f386ff23d4353a5a36&vid=17e0b58af541c85e89fb66f386ff23d4353a5a36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR