4-Hexylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H0890-5G H0890-25G
TCH0890-005GEA 109.02 CAD
TCH0890-005G TCH0890-25G
4-Hexylaniline ≥98.0% (by GC, titration analysis)
4-Hexylaniline

CAS Number: 33228-45-4
MDL Number: MFCD00007927
Molecular Formula: C12H19N
Molecular Weight: 177.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 285
Flash Point (°C): 113
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C

Formula: C₁₂H₁₉N
MW: 177.29 g/mol
Boiling Pt: 285 °C
Density: 0.92 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00007927
CAS Number: 33228-45-4
EINECS: 251-409-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927a9f85db70cabbe1ebbd63d8883a190406b4d1&vid=d656dfd7ddc71e1863c49b702c28f2da60f61460&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR