Hexyltrimethoxysilane ≥98.0%

Supplier: TCI America

H0879-500ML H0879-25ML H0879-100ML
TCH0879-500MLEA 240.16 CAD
TCH0879-500ML TCH0879-025ML TCH0879-100ML
Hexyltrimethoxysilane ≥98.0%
Hexyltrimethoxysilane

CAS Number: 3069-19-0
MDL Number: MFCD00039801
Molecular Formula: C9H22O3Si
Molecular Weight: 206.36
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 203
Flash Point (°C): 77
Specific Gravity (20/20): 0.92

Formula: C₉H₂₂O₃Si
Boiling Pt: 203 °C
Density: 0.92 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00039801
CAS Number: 3069-19-0
EINECS: 221-331-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85f10f90167f2ceb4f5dea1fe272d5afe6617735&vid=85f10f90167f2ceb4f5dea1fe272d5afe6617735&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR