4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98.0% free radical
Supplier: TCI America
Synonyms:
2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, Tempol
CAS Number: 2226-96-2
MDL Number: MFCD00006478
Molecular Formula: C9H18NO2
Molecular Weight: 172.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72
Flash Point (°C): 146
Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Hydroxy-TEMPO
Formula:
C₉H₁₈NO₂ MW: 172.25 g/mol Melting Pt: 72 °C Flash Pt: 146 °C |
MDL Number:
MFCD00006478 CAS Number: 2226-96-2 EINECS: 218-760-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...