(4-Hydroxyphenyl)acetic acid methyl ester ≥98.0%

Supplier: TCI America

Synonyms: Methyl 4-hydroxyphenylacetate, Methyl (4-hydroxyphenyl)acetate, Methyl 2-(4-hydroxyphenyl)acetate

H0856-25G H0856-500G
TCH0856-025GEA 91.64 CAD
TCH0856-025G TCH0856-500G
(4-Hydroxyphenyl)acetic acid methyl ester ≥98.0%
(4-Hydroxyphenyl)acetic acid methyl ester

CAS Number: 14199-15-6
MDL Number: MFCD00002387
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 310
Melting point (°C): 57


Other Product Names / Synonyms: 4-Hydroxyphenylacetic Acid Methyl Ester

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 310 °C
Melting Pt: 57 °C
MDL Number: MFCD00002387
CAS Number: 14199-15-6
EINECS: 238-050-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a8f86983e74823be274e29c364ee2e9ffae65cf&vid=9a8f86983e74823be274e29c364ee2e9ffae65cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR