(4-Hydroxyphenyl)acetamide ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxyphenylacetamide

H0853-25G H0853-500G
TCH0853-025GEA 59.42 CAD
TCH0853-025G TCH0853-500G
(4-Hydroxyphenyl)acetamide ≥98.0%
(4-Hydroxyphenyl)acetamide

CAS Number: 17194-82-0
MDL Number: MFCD00017145
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 177


Other Product Names / Synonyms: 4-Hydroxybenzeneacetamide

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00017145
CAS Number: 17194-82-0
EINECS: 241-235-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62d6fb3fd3cb10077a83cbec9b50db534cc2469b&vid=62d6fb3fd3cb10077a83cbec9b50db534cc2469b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR