7-Hydroxyflavone ≥97.0%

Supplier: TCI America

Synonyms: 7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one

H0852-1G H0852-5G
TCH0852-001GEA 94.17 CAD
TCH0852-001G TCH0852-5G
7-Hydroxyflavone ≥97.0%
7-Hydroxyflavone

CAS Number: 6665-86-7
MDL Number: MFCD00006835
Molecular Formula: C15H10O3
Molecular Weight: 238.24
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 246

Formula: C₁₅H₁₀O₃
Boiling Pt: 395 °C (1013 hPa)
Melting Pt: 246 °C
MDL Number: MFCD00006835
CAS Number: 6665-86-7
EINECS: 229-705-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d05d24e7a8c650236db9459b4a4ceb5bfec4b06&vid=0d05d24e7a8c650236db9459b4a4ceb5bfec4b06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR